
1,3-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:79850-12-7
:
Formula:C₈H₁₀F₆
:220.16 g/mol
Boiling Pt:135°C
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Specification Test Results
Frequently Bought Together