2-(7-Bromo-9,9-di-n-octyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by titrimetric analysis)
Catalogus # TCIAB5580-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:67.0 to 71.0 deg-C
Specification Test Results
About this item
:
:595.51 g/mol
CAS Number:620624-96-6
Formula:C₃₅H₅₂BBrO₂