Order Entry
Netherlands
ContactUsLinkComponent
 
Biotin-PEG4-hydrazide ≥94.0% (by HPLC)
 :  TCI
 
Biotin-PEG4-hydrazide ≥94.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 94.0 area%
  • Elemental analysis (Nitrogen):
    13.00 to 14.50 %

 

Frequently Bought Together