
2,5-Bis(trimethylsilyl)-1,4-phenylene bis(trifluoromethanesulphonate) ≥98.0% (by GC)
Catalog # TCIAB5559-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:105.0 to 109.0 deg-C
Specification Test Results
About this item
:
:518.59 g/mol
UN:3261
ADR:8,II
:GHS05
CAS Number:613676-07-6
Formula:C₁₄H₂₀F₆O₆S₂Si₂