
2-(tert-Butyldimethylsilyl)-1,3-phenylene bis(trifluoromethanesulphonate) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1637638-66-4
:
Formula:C₁₄H₁₈F₆O₆S₂Si
:488.49 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together