
3'-Bromo-2'-methoxyacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:267651-23-0
:
Formula:C₉H₉BrO₂
MDL Number:MFCD22573042
:229.07 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together