Order Entry
Netherlands
ContactUsLinkComponent
4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde ≥97.0% (by GC)
4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde ≥97.0% (by GC)
Catalog # TCIAB5484-200MG
Supplier:  TCI
CAS Number:  
4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde ≥97.0% (by GC)
Catalog # TCIAB5484-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    104.0 to 108.0 deg-C

Specification Test Results

About this item

 :
 :502.73 g/mol
CAS Number:1668554-22-0
Formula:C₂₈H₃₈O₄S₂