
5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:142946-80-3
:
Formula:C₁₂H₆F₆N₂
MDL Number:MFCD24449717
:292.18 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
Specification Test Results
Frequently Bought Together