
3,3'-(Butane-1,4-diyl)bis(1-vinyl-3-imidazolium) bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC)
TCIAB5421-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 96.0 %
- Melting point:80.0 to 84.0 deg-C
:
:804.61 g/mol
:GHS07
:1312310-16-9
:C₁₈H₂₀F₁₂N₆O₈S₄