Order Entry
Netherlands
ContactUsLinkComponent
Bis(1,10-phenanthroline)palladium(II) bis(hexafluorophosphate) ≥98.0% (by titrimetric analysis)
Irritant
Bis(1,10-phenanthroline)palladium(II) bis(hexafluorophosphate) ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
Bis(1,10-phenanthroline)palladium(II) bis(hexafluorophosphate) ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together