Order Entry
Netherlands
ContactUsLinkComponent
 
1-tert-Butyl 5-(N-Succinimidyl) N-Palmitoyl-L-glutamate ≥96.0% (by HPLC)
  TCIAB5390-100MG
 :  TCI
 :  
1-tert-Butyl 5-(N-Succinimidyl) N-Palmitoyl-L-glutamate ≥96.0% (by HPLC)
  TCIAB5390-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Elemental analysis (Nitrogen):
    4.70 to 5.70 %
  • Melting point:
    86.0 to 90.0 deg-C

 

 

 :
 :538.73 g/mol
 :Freezer
 :204521-63-1
 :C₂₉H₅₀N₂O₇
 :MFCD22989465