(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:159622-10-3
:
Formula:C₁₁H₁₇NO₄
MDL Number:MFCD11042412
:227.26 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 96.0 %
- Specific rotation [a]20/D:+20.0 to +24.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together