4,4'''-Bis(2-butyloctyloxy)-p-quaterphenyl ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:18434-08-7
:
Formula:C₄₈H₆₆O₂
MDL Number:MFCD02667784
Melting Pt:163°C
:675.05 g/mol
EINECS:242-310-0
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Molar extinction coefficient:min. 51000 (CHCl3, 308 to 312nm)
Specification Test Results
Frequently Bought Together