
2-Bromo-1-(1-pyrrolidinyl)-1-ethanone ≥93.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90892-09-4
:
Formula:C₆H₁₀BrNO
MDL Number:MFCD06800371
:192.06 g/mol
Boiling Pt:120°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 93.0 %
Specification Test Results
Frequently Bought Together