
(S,S)-(+)-2,3-Bis(tert-butylmethylphosphino)quinoxaline ≥98.0% (by HPLC)
Catalog # TCIAB5302-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):8.00 to 8.80 %
- Melting point:100.0 to 104.0 deg-C
- Specific rotation [a]20/D:+50.0 to +55.0 deg (C=1, CHCL3)
Specification Test Results
About this item
:
:334.38 g/mol
:GHS07
CAS Number:1107608-80-9
Formula:C₁₈H₂₈N₂P₂
MDL Number:MFCD10567042