
(R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline ≥98.0% (by HPLC)
TCIAB5301-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):8.00 to 8.80 %
- Melting point:100.0 to 104.0 deg-C
- Specific rotation [a]20/D:-48.0 to -53.0 deg (C=1, CHCL3)
:
:334.38 g/mol
:GHS07
:866081-62-1
:C₁₈H₂₈N₂P₂
:MFCD10565649