Order Entry
Netherlands
ContactUsLinkComponent
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene ≥95.0% (by GC, titration analysis)
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene ≥95.0% (by GC, titration analysis)
Catalog # TCIAB5288-1G
Supplier:  TCI
CAS Number:  
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene ≥95.0% (by GC, titration analysis)
Catalog # TCIAB5288-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :380.10 g/mol
CAS Number:853377-10-3
Formula:C₂₂H₃₀B₂O₄
MDL Number:MFCD22381082