
1,8-Bis(diphenylphosphinyl)naphthalene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:316808-41-0
:
Formula:C₃₄H₂₆O₂P₂
:528.53 g/mol
:GHS07
Specification Test Results
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together