

(3-Bromophenyl)(mesityl)iodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1203709-76-5
:
Formula:C₁₆H₁₅BrF₃IO₃S
MDL Number:MFCD20264881
:551.16 g/mol
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Ion exchange titration):min. 98.0 %
- Melting point:168.0 to 172.0 deg-C
Specification Test Results
Frequently Bought Together