
(4-Bromobenzyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:51044-13-4
:
Formula:C₂₅H₂₁Br₂P
MDL Number:MFCD00051922
:512.24 g/mol
Melting Pt:275°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together