Bis[2-[(oxo)diphenylphosphino]phenyl] ether ≥97.0% (by HPLC)
Catalog # TCIAB5240-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
Melting Pt:280°C
:570.56 g/mol
CAS Number:808142-23-6
Formula:C₃₆H₂₈O₃P₂
MDL Number:MFCD28968111