4,5-Bis(dicyclohexylphosphino)-9,9-dimethylxanthene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:940934-47-4
:MFCD11559064
Formula:C₃₉H₅₆OP₂
MDL Number:MFCD22422615
:602.82 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:169.0 to 174.0 deg-C
Specification Test Results
Frequently Bought Together