
2-Bromo-4-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
TCIAB5238-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:81.0 to 85.0 deg-C
:
:283.96 g/mol
:GHS07
:458532-82-6
:C₁₁H₁₅BBrNO₂
:MFCD06798252