Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone ≥98.0% (by GC)
 :  TCI
 
undefined
 
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    42.0 to 46.0 deg-C

 

Frequently Bought Together