
(S)-2-(tert-Butoxycarbonyl)isoxazolidine-3-carboxylic acid ≥98.0% (by HPLC)
Catalog # TCIAB5176-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):6.00 to 6.80 %
- Specific rotation [a]20/D:-107 to -117 deg (C=1,CH2CL2)
Specification Test Results
About this item
:
:217.22 g/mol
:GHS07
CAS Number:1372202-46-4
Formula:C₉H₁₅NO₅