
3-(Benzyloxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
TCIAB5172-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 107.0 %
:
:256.06 g/mol
:GHS07
:380430-52-4
:C₁₄H₁₃BO₄
:MFCD02179445