
4-(2-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:101681-34-9
:
Formula:C₁₁H₈BrN
MDL Number:MFCD18415515
:234.10 g/mol
Boiling Pt:89°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:62.0 to 66.0 deg-C
Specification Test Results
Frequently Bought Together