4-Bromo-N,N-bis(9,9-dimethyl-9H-fluoren-2-yl)aniline ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:313050-71-4
:
Formula:C₃₆H₃₀BrN
:556.55 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together