4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:850568-72-8
:
Formula:C₁₇H₂₅BO₄
MDL Number:MFCD03453055
:304.1890 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together