
3,6-Bis(trimethylsilyl)naphthalene-2,7-diyl bis(trifluoromethanesulphonate) ≥98.0% (by GC)
TCIAB5080-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
:
:568.65 g/mol
:56°C
:GHS07
:947488-89-3
:C₁₈H₂₂F₆O₆S₂Si₂