
2-Benzyloxy-5-chlorophenylboronic acid (contains varying amounts of Anhydride)
TCIAB5076-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):95.0 to 107.0 %
:
:262.4966 g/mol
:110°C
:GHS07
:612832-83-4
:C₁₃H₁₂BClO₃
:MFCD04039000