1-n-Butoxy-2,3-difluorobenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:136239-66-2
:
Formula:C₁₀H₁₂F₂O
MDL Number:MFCD07368738
:186.202 g/mol
Boiling Pt:110°C
EINECS:446-810-1
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together