tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate ≥98.0% (by GC, titration analysis)
Catalog # TCIAB5032-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:198.27 g/mol
Storage Temperature:Freezer
CAS Number:1041026-70-3
Formula:C₁₀H₁₈N₂O₂
MDL Number:MFCD11226963