Order Entry
Netherlands
ContactUsLinkComponent
9-(4-Bromophenyl)-10-(2-naphthyl)anthracene ≥97.0% (by HPLC)
9-(4-Bromophenyl)-10-(2-naphthyl)anthracene ≥97.0% (by HPLC)
Supplier:  TCI
Certificates
9-(4-Bromophenyl)-10-(2-naphthyl)anthracene ≥97.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Argentmetric Titration):
    min. 96.0 %

Specification Test Results

Frequently Bought Together