
1-(3-Bromophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:21327-14-0
:
Formula:(C₆H₄Br)NHC(S)NH₂
MDL Number:MFCD00060440
:231.12 g/mol
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:145.0 to 149.0 deg-C
Specification Test Results
Frequently Bought Together