Order Entry
Netherlands
ContactUsLinkComponent
 
N,N'-Bis(9,9-dimethyl-9H-fluoren-2-yl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
  TCIAB4962-1G
 :  TCI
 :  
N,N'-Bis(9,9-dimethyl-9H-fluoren-2-yl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
  TCIAB4962-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

 

 

 :
 :720.96 g/mol
 :361486-60-4
 :C₅₄H₄₄N₂
 :MFCD20528032