
4-Bromo-2-iodophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:207115-22-8
:
Formula:C₆H₄BrIO
MDL Number:MFCD09029644
:298.91 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:72.0 to 76.0 deg-C
Specification Test Results
Frequently Bought Together