2,8-Bis(9H-carbazol-9-yl)dibenzothiophene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:913738-04-2
:
Formula:C₃₆H₂₂N₂S
MDL Number:MFCD28291905
:514.65 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together