
4-(4-Bromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB4933-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:165.0 to 169.0 deg-C
Specification Test Results
About this item
:
:387.28 g/mol
:GHS07
CAS Number:58536-46-2
Formula:C₂₂H₁₅BrN₂
MDL Number:MFCD00474667