
4-(3-Bromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:864377-28-6
:
Formula:C₂₂H₁₅BrN₂
MDL Number:MFCD28962150
:387.28 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:134.0 to 138.0 deg-C
Specification Test Results
Frequently Bought Together