1,3-Benzenedisulphonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:3701-01-7
:
Formula:C₆H₈N₂O₄S₂
MDL Number:MFCD00025383
:236.26 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:233.0 to 237.0 deg-C
Specification Test Results
Frequently Bought Together