trans,trans-4'-(3-Butenyl)-4-(3,4-difluorophenyl)bicyclohexyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:155266-68-5
:
Formula:C₂₂H₃₀F₂
MDL Number:MFCD09839393
:332.48 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:41.0 to 45.0 deg-C
Specification Test Results
Frequently Bought Together