Order Entry
Netherlands
ContactUsLinkComponent
 
trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)
 :  TCI
 
trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    101.0 to 105.0 deg-C

 

Frequently Bought Together