O-tert-Butyl-L-serine tert-butyl ester hydrochloride ≥98.0% (by titrimetric analysis)
: TCI
:51537-21-4
:
:C₁₁H₂₃NO₃·HCl
:MFCD00034850
:253.767 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:-9.0 to -11.0 deg (C=1,H2O)
Frequently Bought Together