N-(tert-Butoxycarbonyl)-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:112525-72-1
:
Formula:C₁₆H₂₀N₂O₄
MDL Number:MFCD00190836
:304.35 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together