2,7-Bis[N-(m-tolyl)anilino]-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1033035-83-4
:
Formula:C₅₁H₃₈N₂
MDL Number:MFCD11112143
:678.88 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:211.0 to 215.0 deg-C
Specification Test Results
Frequently Bought Together