2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:932739-76-9
:
Formula:C₅₇H₃₈N₂
MDL Number:MFCD12022428
:750.95 g/mol
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together