3,3'-Bis[di(p-tolyl)amino]biphenyl ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:161485-60-5
:
Formula:C₄₀H₃₆N₂
MDL Number:MFCD16036283
:544.74 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:191.0 to 195.0 deg-C
Specification Test Results
Frequently Bought Together