

9,9-Bis(4-hydroxyphenyl)fluorene ≥98.0% (by GC), purified by sublimation
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:3236-71-3
:
Formule:C₂₅H₁₈O₂
MDL-nummer:MFCD00191392
:350.42 g/mol
EINECS:406-950-6
UN:3077
ADR:9,III
:GHS07
:GHS09
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:224.0 to 228.0 deg-C
Specificatie testresultaten
Frequently Bought Together