Order Entry
Netherlands
ContactUsLinkComponent
2-Boc-2-azabicyclo[2.2.1]hept-5-ene ≥95.0% (by GC)
Corrosive
2-Boc-2-azabicyclo[2.2.1]hept-5-ene ≥95.0% (by GC)
Catalog # TCIAB4806-1G
Supplier:  TCI
CAS Number:  
Corrosive
2-Boc-2-azabicyclo[2.2.1]hept-5-ene ≥95.0% (by GC)
Catalog # TCIAB4806-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :195.262 g/mol
Boiling Pt:92°C
 :GHS05
CAS Number:188345-71-3
 :MFCD11865216
Formula:C₁₁H₁₇NO₂
MDL Number:MFCD22416354